missing translation for 'onlineSavingsMsg'
Learn More

(2-Hydroxyethyl)hydrazine, Thermo Scientific Chemicals

Product Code. 11310438
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

(2-Hydroxyethyl)hydrazine is used as a plant growth regulant, a component in jet fuels, intermediate for organic synthesis especially heterocycles used in agrochemicals, pharmaceuticals, stabilizers and polymerizations, Chain extender for urethane formulations, corrosion inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-84-2
Molecular Formula C2H8N2O
Molecular Weight (g/mol) 76.099
MDL Number MFCD00007623
InChI Key GBHCABUWWQUMAJ-UHFFFAOYSA-N
Synonym 2-hydroxyethylhydrazine, 2-hydrazinoethanol, omaflora, ethanolhydrazine, 2-hydroxyethyl hydrazine, ethanol, 2-hydrazino, hydrazineethanol, 2-hydrazinoethyl alcohol, beta-hydroxyethylhydrazine, 2-hydrazineethanol
PubChem CID 8017
ChEBI CHEBI:50098
IUPAC Name 2-hydrazinylethanol
SMILES C(CO)NN

Specifica

Melting Point -70°C
Density 1.121 g/mL
Boiling Point 218°C to 220°C
Flash Point 73°C (163°F)
Refractive Index 1.493
Quantity 10 g
UN Number UN2810
Solubility Information Soluble in water,alcohols and glycols.
Formula Weight 76.1
Physical Form Liquid
Chemical Name or Material (2-Hydroxyethyl)hydrazine
Vedi altri risultati Mostra meno risultati

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'