missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-1,4-benzodioxane, 98%, Thermo Scientific Chemicals

Product Code. 11452217
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3663-82-9
Molecular Formula C9H10O3
Molecular Weight (g/mol) 166.176
MDL Number MFCD00006822
InChI Key GWQOQQVKVOOHTI-UHFFFAOYSA-N
Synonym 2-hydroxymethyl-1,4-benzodioxane, 2,3-dihydro-benzo 1,4 dioxin-2-yl-methanol, 1,4-benzodioxan-2-methanol, 2,3-dihydro-1,4-benzodioxin-2-ylmethanol, 2,3-dihydrobenzo b 1,4 dioxin-2-yl methanol, 1,4-benzodioxane-2-methanol, 2-hydroxymethyl-1,4-benzodioxan, 1,4-benzodioxin-2-methanol, 2,3-dihydro
PubChem CID 19314
IUPAC Name 2,3-dihydro-1,4-benzodioxin-3-ylmethanol
SMILES C1C(OC2=CC=CC=C2O1)CO

Specifications

Melting Point 87°C to 91°C
Quantity 5 g
Beilstein 83736
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-Hydroxymethyl-1,4-benzodioxane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.