missing translation for 'onlineSavingsMsg'
Learn More

2-(Hydroxymethyl)tetrahydropyran, 94%, Thermo Scientific Chemicals

Product Code. 15489059
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-72-1
Molecular Formula C6H12O2
Molecular Weight (g/mol) 116.16
MDL Number MFCD00006624
InChI Key ROTONRWJLXYJBD-UHFFFAOYSA-N
Synonym tetrahydropyran-2-methanol, 2h-pyran-2-methanol, tetrahydro, 2-hydroxymethyltetrahydropyran, tetrahydropyran-2-carbinol, 2-hydroxymethyl tetrahydropyran, pyran-2-methanol, tetrahydro, tetrahydro-2h-pyran-2-yl methanol, 2-tetrahydropyranilcarbinol, tetrahydropyranyl-2-methanol, 2-methyloltetrahydro-1,4-pyran
PubChem CID 7524
IUPAC Name oxan-2-ylmethanol
SMILES C1CCOC(C1)CO

Specifications

Melting Point <-70°C
Density 1.027
Boiling Point 186°C to 189°C
Flash Point 93°C (199°F)
Refractive Index 1.458
Quantity 25 g
Beilstein 102998
Formula Weight 116.16
Percent Purity 94%
Chemical Name or Material 2-(Hydroxymethyl)tetrahydropyran
compliance-icons
Product Identifier
  • 2-(Hydroxymethyl)tetrahydropyran
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Tetrahydropyran-2-methanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.