missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethylbenzothiazole, 98%, Thermo Scientific™

Product Code. 10088641
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Spezifikation

CAS 37859-42-0
CAS Min % 97.5
CAS Max % 100.0
Melting Point 95.0°C to 96.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C8H7NOS
MDL Number MFCD00226293
Quantity 1 g
Synonym benzo d thiazol-2-ylmethanol, 2-benzothiazolemethanol, 2-hydroxymethylbenzothiazole, 1,3-benzothiazol-2-yl-methanol, benzothiazol-2-ylmethan-1-ol, zlchem 107, pubchem14535, benzothiazol-2-yl-methanol, chembl71700, 1,3-benzothiazol-2-yl methanol
InChI Key PQXMQZYDBQBWNL-UHFFFAOYSA-N
SMILES OCC1=NC2=CC=CC=C2S1
IUPAC Name 1,3-benzothiazol-2-ylmethanol
Molecular Weight (g/mol) 165.21
PubChem CID 268122
Formula Weight 165.22
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Hydroxymethylbenzothiazole, 98%
Mehr anzeigen Weniger anzeigen
compliance-icons
Produktidentifikator
  • 2-Hydroxymethylbenzothiazole
Signalwort
  • Warning
Gefahrenkategorie
  • Acute toxicity Category 4
Gefahrenhinweis
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Sicherheitshinweis
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'