Learn More
2-Hydroxyphenylacetic acid, 98%, Thermo Scientific Chemicals
37.39 € - 123.62 €
Chemical Identifiers
CAS | 614-75-5 |
---|---|
Molecular Formula | C8H8O3 |
Molecular Weight (g/mol) | 152.149 |
MDL Number | MFCD00004323 |
InChI Key | CCVYRRGZDBSHFU-UHFFFAOYSA-N |
Synonym | 2-hydroxyphenylacetic acid, 2-2-hydroxyphenyl acetic acid, 2-hydroxyphenyl acetic acid, benzeneacetic acid, 2-hydroxy, 2-hydroxybenzeneacetic acid, o-hydroxyphenylacetic acid, acetic acid, o-hydroxyphenyl, 2-hydroxyphenylacetate, o-hydroxyphenyl acetic acid, ortho-hydroxyphenylacetic acid |
PubChem CID | 11970 |
ChEBI | CHEBI:28478 |
IUPAC Name | 2-(2-hydroxyphenyl)acetic acid |
SMILES | C1=CC=C(C(=C1)CC(=O)O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11431648
|
Thermo Scientific Alfa Aesar
A13924.09 |
10 g |
37.39 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11441648
|
Thermo Scientific Alfa Aesar
A13924.18 |
50 g |
123.62 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Hydroxyphenylacetic Acid is an intermediate in the synthesis of atenolol and 3,4-dihydroxyphenylacetic acid.
Solubility
Soluble in Methanol and Water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
614-75-5 | |
152.149 | |
CCVYRRGZDBSHFU-UHFFFAOYSA-N | |
11970 | |
2-(2-hydroxyphenyl)acetic acid |
C8H8O3 | |
MFCD00004323 | |
2-hydroxyphenylacetic acid, 2-2-hydroxyphenyl acetic acid, 2-hydroxyphenyl acetic acid, benzeneacetic acid, 2-hydroxy, 2-hydroxybenzeneacetic acid, o-hydroxyphenylacetic acid, acetic acid, o-hydroxyphenyl, 2-hydroxyphenylacetate, o-hydroxyphenyl acetic acid, ortho-hydroxyphenylacetic acid | |
CHEBI:28478 | |
C1=CC=C(C(=C1)CC(=O)O)O |
Specifications
614-75-5 | |
C8H8O3 | |
908000 | |
Soluble in Methanol and Water. | |
C1=CC=C(C(=C1)CC(=O)O)O | |
152.149 | |
CHEBI:28478 | |
98% |
145°C to 147°C | |
MFCD00004323 | |
2-hydroxyphenylacetic acid, 2-2-hydroxyphenyl acetic acid, 2-hydroxyphenyl acetic acid, benzeneacetic acid, 2-hydroxy, 2-hydroxybenzeneacetic acid, o-hydroxyphenylacetic acid, acetic acid, o-hydroxyphenyl, 2-hydroxyphenylacetate, o-hydroxyphenyl acetic acid, ortho-hydroxyphenylacetic acid | |
CCVYRRGZDBSHFU-UHFFFAOYSA-N | |
2-(2-hydroxyphenyl)acetic acid | |
11970 | |
152.15 | |
2-Hydroxyphenylacetic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 210-393-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.