missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Imidazolidone, 96%, anhydrous, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Melting Point | 131.4°C to 132.4°C |
Color | White |
Boiling Point | 187.0°C (10.0 Torr) |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 250 g |
Beilstein | 24,2 |
Show More |
Safety and Handling
Product Identifier
- 2-Imidazolidone, anhydrous
- Warning
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity after repeated exposure Category 2
- H319-Causes serious eye irritation.
- H373-May cause damage to organs.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P314-Get medical advice/attention if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 2-Imidazolidinone
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'