Learn More
2-Indanone, 98%, Thermo Scientific Chemicals
50.80 € - 604.00 €
Chemical Identifiers
CAS | 615-13-4 |
---|---|
Molecular Formula | C9H8O |
Molecular Weight (g/mol) | 132.162 |
MDL Number | MFCD00003792 |
InChI Key | UMJJFEIKYGFCAT-UHFFFAOYSA-N |
Synonym | 2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone |
PubChem CID | 11983 |
ChEBI | CHEBI:27930 |
IUPAC Name | 1,3-dihydroinden-2-one |
SMILES | C1C(=O)CC2=CC=CC=C21 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11474274
|
Thermo Scientific Alfa Aesar
A12574.06 |
5 g |
50.80 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11494274
|
Thermo Scientific Alfa Aesar
A12574.22 |
100 g | |||||||
Description
2-Indanone undergoes TiCl4-Mg mediated coupling with CHBr3 to yield dibromomethyl carbinol. It reacts with 5,5-dimethyl-3-pyrazolidinone to yield 5,5-dimethyl-2-(1H-indenyl-2)-3-pyrazolidinone. On photolysis by 266-nm one-photon excitation yields o-xylylene. It was used as starting reagent in the synthesis of indene-fused porphyrins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Indanone undergoes TiCl4-Mg mediated coupling with CHBr3 to yield dibromomethyl carbinol. It reacts with 5,5-dimethyl-3-pyrazolidinone to yield 5,5-dimethyl-2-(1H-indenyl-2)-3-pyrazolidinone. On photolysis by 266-nm one-photon excitation yields o-xylylene. It was used as starting reagent in the synthesis of indene-fused porphyrins.
Solubility
Insoluble in water.
Notes
Hygroscopic. Incompatible with oxidizing agents. Incompatible with oxidizing agents and bases.
Chemical Identifiers
615-13-4 | |
132.162 | |
UMJJFEIKYGFCAT-UHFFFAOYSA-N | |
11983 | |
1,3-dihydroinden-2-one |
C9H8O | |
MFCD00003792 | |
2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone | |
CHEBI:27930 | |
C1C(=O)CC2=CC=CC=C21 |
Specifications
615-13-4 | |
218°C (decomposition) | |
C9H8O | |
636550 | |
Insoluble in water. | |
C1C(=O)CC2=CC=CC=C21 | |
132.162 | |
CHEBI:27930 | |
98% |
52°C to 56°C | |
100°C (212°F) | |
MFCD00003792 | |
2-indanone, indan-2-one, 1h-inden-2 3h-one, 1,3-dihydro-2h-inden-2-one, beta-hydrindone, 2h-inden-2-one, 1,3-dihydro, unii-0i79n673de, 2,3-dihydro-1h-inden-2-one, 4-07-00-01002 beilstein handbook reference, 2-lndanone | |
UMJJFEIKYGFCAT-UHFFFAOYSA-N | |
1,3-dihydroinden-2-one | |
11983 | |
132.16 | |
2-Indanone |
Safety and Handling
missing translation for 'einecsNumber' : 210-410-3
missing translation for 'rtecsNumber' : NK7535500
missing translation for 'tsca' : Yes
Recommended Storage : Keep cold; Store under Nitrogen
RUO – Research Use Only