missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-2-methylpropane, 95%, stab. with copper powder, Thermo Scientific Chemicals

Product Code. 11317845
Click to view available options
Quantity:
25 g
100 g

2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Iodo-2-methylpropane is used as a chemical reagent in organic synthesis. Further, it serves as a pharmaceutical intermediate.

Solubility
Miscible with ethanol and ether.

Notes
Light sensitive. Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 558-17-8
Molecular Formula C4H9I
Molecular Weight (g/mol) 184.02
MDL Number MFCD00001068
InChI Key ANGGPYSFTXVERY-UHFFFAOYSA-N
Synonym tert-butyl iodide, propane, 2-iodo-2-methyl, trimethyliodomethane, t-butyl iodide, unii-e5hy19g10p, 2-jod-2-methylpropan, sjphavimup@, tert-c4h9i, 2-iodo-2-methyl propane, 2-iodo-2-methyl-propane
PubChem CID 11206
IUPAC Name 2-iodo-2-methylpropane
SMILES CC(C)(C)I

Specifications

Melting Point -38°C
Density 1.547
Boiling Point 99°C to 100°C
Flash Point 7°C (44°F)
Odor Recognizable
Refractive Index 1.491
Quantity 25 g
UN Number UN2391
Beilstein 505950
Sensitivity Light sensitive
Solubility Information Miscible with ethanol and ether.
Formula Weight 184.02
Percent Purity 95%
Chemical Name or Material 2-Iodo-2-methylpropane, stabilized with copper powder
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.