missing translation for 'onlineSavingsMsg'
Learn More
2-Iodoethanol, 99%, stabilized, Thermo Scientific Chemicals Product Code.: 10561431

2-Iodoethanol, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10561431
50 mL
Click to view available options
Quantity:
10 mL
50 mL
250 mL
This item is not returnable. View return policy

Product Code. 10561431

Brand: Thermo Scientific Acros 175500500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 624-76-0
Molecular Formula C2H5IO
Molecular Weight (g/mol) 171.97
MDL Number MFCD00002830
InChI Key QSECPQCFCWVBKM-UHFFFAOYSA-N
Synonym ethanol, 2-iodo, iodoethanol, ethylene iodohydrin, 2-iodoethan-1-ol, iodoethyl alcohol, 2-indoethanol, 2-iodo ethanol, 2-iodo-ethanol, 2-iodo-1-ethanol, 2-iodoethyl alcoholShow MoreShow Less
PubChem CID 12225
SMILES OCCI

Specifications

Density 2.2050g/mL
Boiling Point 172.0°C to 175.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ICH2CH2OH
Refractive Index 1.5690 to 1.5720
Quantity 50 mL
Beilstein 01,339
Fieser 13,348
Specific Gravity 2.205
IUPAC Name 2-iodoethanol
Stabilizer Copper
Formula Weight 171.97
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Iodoethanol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467902 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number175500100,175500500
Lot NumberA0463682 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number175500100,175500500
Lot NumberA0463124 Certificate TypeCertificate of Analysis Date24/05/2024 Catalog Number175500100,175500500
Lot NumberA0397969 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number175500100,175502500,175500500
Lot NumberA0374386 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number175500100,175502500,175500500
compliance-icons
Product Identifier
  • 2-Iodoethanol, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Iodoethanol, 99%, stabilized, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.