missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isocyanatobenzonitrile, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Linear Formula | CNC6H4NCO |
Quantity | 250 mg |
Synonym | 2-cyanophenyl isocyanate, benzonitrile,2-isocyanato, 2-isocyanato-benzonitrile, 2-isocyanatobenzenecarbonitrile, 2-cyanophenylisocyanate, benzonitrile, 2-isocyanato, acmc-1ap1b |
Formula Weight | 144.13 |
Percent Purity | 95% |
Chemical Name or Material | 2-Isocyanatobenzonitrile |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'