missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropoxybenzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 15499927
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138008-97-6
Molecular Formula C9H13BO3
Molecular Weight (g/mol) 180.01
MDL Number MFCD03427050
InChI Key XDMKBIIRBDPSOE-UHFFFAOYSA-N
Synonym 2-isopropoxyphenylboronic acid, 2-isopropoxyphenyl boronic acid, 2-isopoxybenzeneboronic acid, 2-isopropoxybenzeneboronic acid, boronic acid, 2-1-methylethoxy phenyl, 2-propan-2-yloxy phenylboronic acid, 2-isopropoxyphenylboronicacid, 2-propan-2-yloxyphenyl boronic acid, akos brn-0366, pubchem13297
PubChem CID 5084099
IUPAC Name (2-propan-2-yloxyphenyl)boronic acid
SMILES CC(C)OC1=CC=CC=C1B(O)O

Specifications

Melting Point 30°C to 35°C
Density 1.075
Boiling Point 199°C
Flash Point 110°C (230°F)
Refractive Index 1.509
Quantity 1 g
Formula Weight 180.01
Percent Purity 95%
Chemical Name or Material 2-Isopropoxybenzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'