missing translation for 'onlineSavingsMsg'
Learn More

2-Isopropyl-4-methylthiazole, 98%, Thermo Scientific™

Product Code. 11455643
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15679-13-7
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.232
MDL Number MFCD00036561
InChI Key OFLXNHNYPQPQKW-UHFFFAOYSA-N
Synonym 2-isopropyl-4-methylthiazole, 4-methyl-2-1-methylethyl thiazole, 2-isopropyl-4-methyl thiazole, thiazole, 2-isopropyl-4-methyl, thiazole, 4-methyl-2-1-methylethyl, unii-nqf7jo3mup, tropical thiazole, 2-isopropyl-4-methyl-1,3-thiazole, fema no. 3555, nqf7jo3mup
PubChem CID 61808
IUPAC Name 4-methyl-2-propan-2-yl-1,3-thiazole
SMILES CC1=CSC(=N1)C(C)C

Specifications

Density 1
Boiling Point 174°C to 176°C
Flash Point 60°C (140°F)
Odor Characteristic
Refractive Index 1.5
Quantity 5 g
UN Number UN1993
Beilstein 1209894
Formula Weight 141.24
Percent Purity 98%
Chemical Name or Material 2-Isopropyl-4-methylthiazole
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'