missing translation for 'onlineSavingsMsg'
Learn More

2-Mercapto-4,5-diphenyloxazole, 98%, Thermo Scientific™

Product Code. 11410631
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6670-13-9
Molecular Formula C15H11NOS
Molecular Weight (g/mol) 253.319
MDL Number MFCD00173660
InChI Key NYQSTRUSYDZNHC-UHFFFAOYSA-N
Synonym 4,5-diphenyl-2-oxazolethiol, 4,5-diphenyl-1,3-oxazole-2-thiol, diphenyl-1,3-oxazole-2-thiol, 4,5-diphenyl-2-mercaptooxazole, 4,5-diphenyl-1,3-oxazole-2 3h-thione, 4,5-diphenyloxazole-2 3h-thione, 4,5-diphenyl-4-oxazoline-2-thione, 4,5-diphenyl-2,3-dihydro-1,3-oxazole-2-thione, 4,5-diphenyloxazole-2-thiol, 4,5-diphenyl-3h-oxazole-2-thione
PubChem CID 853676
IUPAC Name 4,5-diphenyl-3H-1,3-oxazole-2-thione
SMILES C1=CC=C(C=C1)C2=C(OC(=S)N2)C3=CC=CC=C3

Specifications

Melting Point 251°C to 256°C
Quantity 5 g
Sensitivity Air Sensitive
Formula Weight 253.33
Percent Purity 98%
Chemical Name or Material 2-Mercapto-4,5-diphenyloxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'