Learn More
2-Mercapto-5-methylbenzimidazole, 98%, Thermo Scientific Chemicals
Description
2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Mercapto-5-methylbenzimidazole is used as an additive in liquid paraffin. And also used in electrochemical techniques, corrosion inhibitors for steel under highly acidic conditions and pointed out that the effectiveness of the heterocyclic compound can be intimately associated to its molecular structure.
Solubility
Insoluble in water.
Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Specifications
Specifications
Melting Point | ∼295°C (decomposition) |
Quantity | 5 g |
Beilstein | 5083 |
Solubility Information | Insoluble in water. |
Formula Weight | 164.23 |
Percent Purity | 98% |
Chemical Name or Material | 2-Mercapto-5-methylbenzimidazole |
Safety and Handling

- 2-Mercapto-5-methylbenzimidazole
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 2-Mercapto-5-methylb
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.