missing translation for 'onlineSavingsMsg'
Learn More
2-Mercaptobenzoxazole, 99%, Thermo Scientific Chemicals Product Code.: 10285170

2-Mercaptobenzoxazole, 99%, Thermo Scientific Chemicals

Product Code. 10285170
25 g, Glass bottle
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10285170

Brand: Thermo Scientific Acros 125460250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2382-96-9
CAS Min % 98.5
CAS Max % 100.0
Melting Point 192°C to 195°C
Color Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Molecular Formula C7H5NOS
MDL Number MFCD00005769
Quantity 25 g
Beilstein 27, 181
Synonym 2-mercaptobenzoxazole, benzo d oxazole-2-thiol, 2-benzoxazolethiol, 2 3h-benzoxazolethione, benzoxazole-2-thiol, 2-benzoxazolinethione, 1,3-benzoxazole-2-thiol, benzoxazolinethione, 2-mercapto-1,3-benzoxazole, benzooxazole-2-thiol
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in aqueous alkali
InChI Key FLFWJIBUZQARMD-UHFFFAOYSA-N
SMILES C1=CC=C2C(=C1)NC(=S)O2
IUPAC Name 3H-1,3-benzoxazole-2-thione
Molecular Weight (g/mol) 151.18
PubChem CID 712377
Formula Weight 151.18
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Mercaptobenzoxazole, 99%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0379597 Certificate TypeCertificate of Analysis Date13/12/2021 Catalog Number125460250

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Mercaptobenzoxazole, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.