missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ 2-Mercaptoethanol (50 mM)

2-Mercaptoethanol (50 mM)

Brand:  Gibco™ 31350010


Product Code. 11528926

  • 13.76 € / 20mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Gibco™ 2-Mercaptoethanol (also known as beta-mercaptoethanol or BME) is a potent reducing agent used in cell culture medium to prevent toxic levels of oxygen radicals. Mercaptoethanol is not stable in solution so most protocols require daily supplementation. Gibco™ 2-Mercaptoethanol is made at 50 mM in Dulbecco's Phosphate Buffered Saline (DPBS).

Product Use
For Research Use Only. Not for use in diagnostic procedures.

cGMP Manufacturing and Quality System
Gibco™ 2-Mercaptoethanol is manufactured at a cGMP compliant facility, located in Paisley, Scotland, UK. The facility is registered with the FDA as a medical device manufacturer and is certified to the ISO 13485 standard.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: 2 – 8°C
Shipping conditions: Ambient
Shelf life: 36 months from date of manufacture
50 mM
BME
20 mL
Mammalian Cells
Liquid
6 to 8
Room Temperature
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 2-mercaptoethanol

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P273-Avoid release to the environment.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : 2-Mercaptoethanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.