missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptoethyl sulfide, 90%, Thermo Scientific Chemicals

Product Code. 12664937
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3570-55-6
Molecular Formula C4H10S3
Molecular Weight (g/mol) 154.30
MDL Number MFCD00004893
InChI Key KSJBMDCFYZKAFH-UHFFFAOYSA-N
Synonym 2,2'-thiodiethanethiol, bis 2-mercaptoethyl sulfide, 2-mercaptoethyl sulfide, ethanethiol, 2,2'-thiobis, bis mercaptoethyl sulfide, 3-thia-1,5-pentanedithiol, 2-mercaptoethylsulfide, 2,2'-dimercaptodiethyl sulfide, unii-oeu4azc07s, ethanethiol, 2,2'-thiodi
PubChem CID 77117
IUPAC Name 2-(2-sulfanylethylsulfanyl)ethanethiol
SMILES SCCSCCS

Specifications

Melting Point -11.0°C
Color Colorless
Density 1.1830g/mL
Boiling Point 135.0°C to 136.0°C (10.0 mmHg)
Flash Point 90°C
Infrared Spectrum Authentic
Assay Percent Range 88 % min. (GC)
Packaging Glass bottle
Linear Formula (HSCH2CH2)2S
Refractive Index 1.594
Quantity 10 g
Specific Gravity 1.183
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,toluene,chloroform,,dioxane,carbon tetrachloride
Formula Weight 154.32
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 2-Mercaptoethyl sulfide
Show More Show Less
Product Identifier
  • 2-Mercaptoethyl sulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.