missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxy-6-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™

Product Code. 15434897
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Used as catalyst in diethylzinc addition to aldehydes.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017778-98-1
Molecular Formula C9H7F3O2
Molecular Weight (g/mol) 204.15
MDL Number MFCD09832324
InChI Key LLBTYKHVRXDHKF-UHFFFAOYSA-N
Synonym 2-methoxy-6-trifluoromethyl benzaldehyde, benzaldehyde, 2-methoxy-6-trifluoromethyl, 6-methoxy-2-trifluoromethyl benzaldehyde, 2-methoxy-6-trifluoromethyl benzaldehyde, jrd
PubChem CID 20111744
IUPAC Name 2-methoxy-6-(trifluoromethyl)benzaldehyde
SMILES COC1=CC=CC(=C1C=O)C(F)(F)F

Specifications

Melting Point 46°C to 49°C
Quantity 1 g
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 204.15
Percent Purity 97%
Chemical Name or Material 2-Methoxy-6-(trifluoromethyl)benzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'