missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-6-(trifluoromethyl)benzaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Used as catalyst in diethylzinc addition to aldehydes.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Used as catalyst in diethylzinc addition to aldehydes.
Solubility
Insoluble in water.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air.
Specifications
Melting Point | 46°C to 49°C |
Quantity | 5 g |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 204.15 |
Percent Purity | 97% |
Chemical Name or Material | 2-Methoxy-6-(trifluoromethyl)benzaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'