missing translation for 'onlineSavingsMsg'
Learn More

2'-Methoxyacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 11433914
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 579-74-8
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD00008725
InChI Key DWPLEOPKBWNPQV-UHFFFAOYSA-N
Synonym 2'-methoxyacetophenone, 1-2-methoxyphenyl ethanone, o-methoxyacetophenone, ethanone, 1-2-methoxyphenyl, o-acetylanisole, 1-2-methoxyphenyl ethan-1-one, o-acetanisole, 2-acetylanisole, 2-methyoxyacetophenone oxime, ortho-methoxyacetophenone
PubChem CID 68481
IUPAC Name 1-(2-methoxyphenyl)ethanone
SMILES COC1=CC=CC=C1C(C)=O

Specifications

Density 1.09
Boiling Point 119°C to 120°C (10 mmHg)
Flash Point 108°C (226°F)
Refractive Index 1.5393
Quantity 25 g
Beilstein 1100602
Sensitivity Light sensitive
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 2'-Methoxyacetophenone
compliance-icons
Product Identifier
  • 2'-Methoxyacetophenone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 2'-Methoxyacetophenone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'