missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxyethanol, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15568814
Click to view available options
Quantity:
500 mL
Packaging:
Narrow Mouth Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 109-86-4
Flash Point 39.00°C
Packaging Narrow Mouth Amber Glass Bottle
Molecular Formula C3H8O2
Quantity 500 mL
Specific Gravity 1L = 0.96kg
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
SMILES COCCO
IUPAC Name 2-methoxyethanol
Molecular Weight (g/mol) 76.095
PubChem CID 8019
ChEBI CHEBI:46790
Formula Weight 76.1
Grade BAKER ANALYZED™ ACS Reagent
Chemical Name or Material 2-Methoxyethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 3
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.