missing translation for 'onlineSavingsMsg'
Learn More

2-Methoxypropene, 95%, stab. with ca 0.5% potassium carbonate, Thermo Scientific Chemicals

Product Code. 11411087
Click to view available options
Quantity:
50 g
250 g

It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for Synthesis of heterocycle compounds and an important intermediate of medicine.

Solubility
Insoluble in water.

Notes
Light sensitive. Store in a cool, dry place in tightly closed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 116-11-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD00014929
InChI Key YOWQWFMSQCOSBA-UHFFFAOYSA-N
Synonym 2-methoxypropene, isopropenyl methyl ether, 1-propene, 2-methoxy, 2-methoxy-1-propene, methyl isopropenyl ether, ether, isopropenyl methyl, propene, 2-methoxy, unii-15wbg0jt6b, 2-methoxy propene, 15wbg0jt6b
PubChem CID 8300
IUPAC Name 2-methoxyprop-1-ene
SMILES CC(=C)OC

Specifications

Density 0.758
Boiling Point 35°C to 36°C
Flash Point −29°C (−20°F)
Odor Ether-like
Refractive Index 1.3835
Quantity 250 g
UN Number UN1993
Beilstein 1734635
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 72.11
Percent Purity 95%
Chemical Name or Material 2-Methoxypropene, stabilized with ≈0.5% potassium carbonate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.