missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-1,3-benzoxazol-4-amine, 97%, Thermo Scientific™ Product Code.: 10698160

2-Methyl-1,3-benzoxazol-4-amine, 97%, Thermo Scientific™

Product Code. 10698160
1 g
Click to view available options
Quantity:
1 g
This item is not returnable. View return policy

Product Code. 10698160

Brand: Thermo Scientific Maybridge MAY00305DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 342897-54-5
Assay Percent Range 97%
Molecular Formula C8H8N2O
Quantity 1 g
Synonym 2-methylbenzo d oxazol-4-amine, 4-amino-2-methyl-1,3-benzoxazole, methylbenzoxazolamine, 2-methyl-4-benzoxazolamine, 2-methylbenzoxazol-4-amine, 2-methyl-4-aminobenzoxazole, 2-methylbenzoxazole-4-amine, 4-amino-2-methylbenzoxazole, 2-methylbenzoxazole-4-ylamine, 4-benzoxazolamine, 2-methyl
InChI Key OEULNBJKLTXMSN-UHFFFAOYSA-N
SMILES CC1=NC2=C(C=CC=C2O1)N
IUPAC Name 2-methyl-1,3-benzoxazol-4-amine
Molecular Weight (g/mol) 148.165
PubChem CID 10606897
Formula Weight 148.16
Chemical Name or Material 2-Methyl-1, 3-benzoxazol-4-amine
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-1,3-benzoxazol-4-amine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.