missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-1,3-dithiolane, 99%, Thermo Scientific Chemicals Product Code.: 11470977

2-Methyl-1,3-dithiolane, 99%, Thermo Scientific Chemicals

Product Code. 11470977
25 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11470977

Brand: Thermo Scientific Alfa Aesar B23372.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5616-51-3
Molecular Formula C4H8S2
Molecular Weight (g/mol) 120.23
MDL Number MFCD01729666
InChI Key CARJCVDELAMAEJ-UHFFFAOYSA-N
Synonym 1,3-dithiolane, 2-methyl, 2-methyl-1,3-dithiacyclopentane, unii-4o81r0spr0, 1,3-dithiolane,2-methyl, fema no. 3705, acmc-20anre, acetaldehdye ethylene thioacetalShow MoreShow Less
PubChem CID 21828
IUPAC Name 2-methyl-1,3-dithiolane
SMILES CC1SCCS1

Specifications

Density 1.15
Boiling Point 40°C (2 mmHg)
Flash Point >100°C (212°F)
Odor Unpleasant
Quantity 25 g
Formula Weight 120.23
Percent Purity 99%
Chemical Name or Material 2-Methyl-1,3-dithiolane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10240867 Certificate TypeCertificate of Analysis Date24/05/2022 Catalog NumberB23372.14,B23372.06
Lot Number10220706 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23372.14,B23372.06
Lot Number10231619 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23372.14,B23372.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-1,3-dithiolane, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.