missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-1-butene, 98%, Thermo Scientific Chemicals Product Code.: 11352229

2-Methyl-1-butene, 98%, Thermo Scientific Chemicals

Product Code. 11352229
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11352229

Brand: Thermo Scientific Alfa Aesar H27230.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Methyl-1-butene is used as a solvent in organic synthesis. It is also used in the preparation of pinacolone, flavor enhancer, spice, crop protectants and tertiary amyl phenol. Further, it serves as a photosensitive material and concrete dispersant. In addition to this, it is used as a fuel and fuel additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-butene is used as a solvent in organic synthesis. It is also used in the preparation of pinacolone, flavor enhancer, spice, crop protectants and tertiary amyl phenol. Further, it serves as a photosensitive material and concrete dispersant. In addition to this, it is used as a fuel and fuel additive.

Solubility
Soluble in ether, ethanol and benzene. Insoluble in water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 563-46-2
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009333
InChI Key MHNNAWXXUZQSNM-UHFFFAOYSA-N
Synonym 2-methyl-1-butene, 2-methylbutene, 1-butene, 2-methyl, 1-isoamylene, 2-methylbutene-1, butene, 2-methyl, gamma-isoamylene, .gamma.-isoamylene, unii-33c9y0i55h, c2h5c ch3 =ch2
PubChem CID 11240
ChEBI CHEBI:77915
IUPAC Name 2-methylbut-1-ene
SMILES CCC(C)=C

Specifications

Melting Point -137°C
Density 0.651
Boiling Point 31°C to 32°C
Flash Point −48°C (−54°F)
Refractive Index 1.374
Quantity 5 g
UN Number UN2459
Solubility Information Soluble in ether,ethanol and benzene. Insoluble in water.
Formula Weight 70.13
Percent Purity 98%
Chemical Name or Material 2-Methyl-1-butene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249400 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog NumberH27230.14,H27230.06
Lot Number10245877 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberH27230.14,H27230.06
Lot NumberS10F048 Certificate TypeCertificate of Analysis Date25/07/2022 Catalog NumberH27230.14,H27230.06
Lot NumberS27I029 Certificate TypeCertificate of Analysis Date01/07/2022 Catalog NumberH27230.14,H27230.06
Lot NumberU19C045 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH27230.14,H27230.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-1-butene, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.