missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-1-phenyl-2-propanol, 98+%, Thermo Scientific Chemicals

Product Code. 11392256
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methyl-1-phenyl-2-propanol is used in confecting floral essences, such as lily, narcissus, jasmine and keiskei and other advanced floral essences. Its acetic ester has fresh fragrance so that it has especial value.

Solubility
Slightly soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-86-7
Molecular Formula C10H14O
Molecular Weight (g/mol) 150.221
MDL Number MFCD00004465
InChI Key RIWRBSMFKVOJMN-UHFFFAOYSA-N
Synonym 2-methyl-1-phenyl-2-propanol, benzyldimethylcarbinol, 2-benzyl-2-propanol, phenyl-tert-butanol, dimethylbenzylcarbinol, dmbc, 1,1-dimethylphenylethanol, 1,1-dimethyl-2-phenylethanol, benzylpropyl alcohol, dimethyl benzyl carbinol
PubChem CID 7531
IUPAC Name 2-methyl-1-phenylpropan-2-ol
SMILES CC(C)(CC1=CC=CC=C1)O

Specifications

Melting Point 23°C to 25°C
Density 0.974
Boiling Point 215°C
Flash Point 82°C (179°F)
Refractive Index 1.515
Quantity 500 g
Beilstein 1855608
Solubility Information Slightly soluble in water.
Formula Weight 150.22
Percent Purity ≥98%
Chemical Name or Material 2-Methyl-1-phenyl-2-propanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.