missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butanol, 99%, pure, Thermo Scientific Chemicals

97.70 € - 467.00 €

Chemical Identifiers

CAS 75-85-4
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00004478
InChI Key MSXVEPNJUHWQHW-UHFFFAOYSA-N
Synonym 2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
PubChem CID 6405
IUPAC Name 2-methylbutan-2-ol
SMILES CCC(C)(C)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10498910
View Documents Promotion Details
Thermo Scientific Acros
166620010
1 L
97.70 €
1 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10062180
View Documents Promotion Details
Thermo Scientific Acros
166620025
2.5 L
208.00 €
2.50 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
10439573
View Documents Promotion Details
Thermo Scientific Acros
166620100
10 L
467.00 €
10 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

75-85-4
88.15
MSXVEPNJUHWQHW-UHFFFAOYSA-N
6405
CCC(C)(C)O
C5H12O
MFCD00004478
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
2-methylbutan-2-ol

Specifications

75-85-4
Colorless
102.0°C
Authentic
Glass Bottle
C2H5C(CH3)2OH
MFCD00004478
15,7253
2-methyl-2-butanol, tert-amyl alcohol, amylene hydrate, tert-pentyl alcohol, tert-pentanol, t-amyl alcohol, 2-butanol, 2-methyl, dimethylethylcarbinol, ethyldimethylcarbinol, tert-isoamyl alcohol
MSXVEPNJUHWQHW-UHFFFAOYSA-N
2-methylbutan-2-ol
6405
88.15
Pure
2-Methyl-2-butanol, 99%
-12.0°C
0.8000g/mL
20°C
98.5% min. (GC)
C5H12O
1.4040 to 1.4060
01,388
0.8
Solubility in water: 70.7g/L (25°C). Other solubilities: miscible with alcohol,ether,benzene,chloroform,,glycerol,oils and acetone
CCC(C)(C)O
88.15
3.7 mPa.s (25°C)
99%
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Harmful if inhaled.
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful in contact with skin.
May cause drowsiness or d

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 200-908-9

missing translation for 'rtecsNumber' : SC0175000

missing translation for 'tsca' : TSCA

Product Identifier
  • 2-Methyl-2-butanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only