missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-butene, 99+%, Thermo Scientific Chemicals

Product Code. 10646902
Click to view available options
Quantity:
25 mL
250 mL
Packaging:
Glass bottle

2-Methyl-2-butene, 99+%, C5H10, CAS Number-513-35-9, 1,1,2-trimethylethylene, 2-butene, 2-methyl, 2-methyl-2-butene, 2-methylbutene-2, 3-methyl-2-butene, amylene, beta-isoamylene, ethylene, trimethyl, n-amylene, trimethylethylene, 250mL, 99% min. (GC), 01, 211, 35.0 deg.C to 38.0 deg.C, 100.0, 99.0

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 513-35-9
Molecular Formula C5H10
Molecular Weight (g/mol) 70.14
MDL Number MFCD00009276
InChI Key BKOOMYPCSUNDGP-UHFFFAOYSA-N
Synonym 2-methyl-2-butene, 2-butene, 2-methyl, trimethylethylene, amylene, n-amylene, 3-methyl-2-butene, ethylene, trimethyl, beta-isoamylene, 1,1,2-trimethylethylene, 2-methylbutene-2
PubChem CID 10553
ChEBI CHEBI:77916
IUPAC Name 2-methylbut-2-ene
SMILES CC=C(C)C

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -134.0°C
Color Colorless to Yellow
Density 0.6600g/mL
Boiling Point 35.0°C to 38.0°C
Flash Point −45°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH=C(CH3)2
Refractive Index 1.3860 to 1.3880
Quantity 25 mL
Beilstein 01, 211
Merck Index 15, 601
Specific Gravity 0.66
Formula Weight 70.13
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-butene, 99+%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'type03FormHeader'

missing translation for 'productTitle'

missing translation for 'thankYou' missing translation for 'feedbackSubmitted'