missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-2-oxazoline, 99%

Product Code. 10437190 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10437190

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1120-64-5 | C4H7NO | 85.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-64-5
Molecular Formula C4H7NO
Molecular Weight (g/mol) 85.11
MDL Number MFCD00005298
InChI Key GUXJXWKCUUWCLX-UHFFFAOYSA-N
Synonym 2-methyl-2-oxazoline, 2-methyloxazoline, 2-methyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-2-methyl, 2-methyl-2-oxazolin, 2-methyl-1,3-oxazoline, 2-oxazoline, 2-methyl, 4,5-dihydro-2-methyl-1,3-oxazole, poly 2-methyl-2-oxazoline, methyl oxazoline
PubChem CID 70713
ChEBI CHEBI:53614
IUPAC Name 2-methyl-4,5-dihydro-1,3-oxazole
SMILES CC1=NCCO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0000g/mL
Boiling Point 109.4°C to 110.4°C
Flash Point 20°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Refractive Index 1.4330 to 1.4350
Quantity 500 g
Beilstein 27, 13
Specific Gravity 1
Formula Weight 85.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methyl-2-oxazoline, 99%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Methyl-2-oxazoline
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H225-Highly flammable liquid and vapour.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.