missing translation for 'onlineSavingsMsg'
Learn More
2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals Product Code.: 11489386

2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals

Product Code. 11489386
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11489386

Brand: Thermo Scientific Alfa Aesar A19090.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to study its antagonist activity. It reacts with a second target site to alter the spatial relation between the hydrophobic agonist binding site and the allosteric site. It may initiate the polymerization of isocyanates and epoxides.

Solubility
Soluble in water at 20°C 32g/L. Soluble in alcohol.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 590-36-3
Molecular Formula C6H14O
Molecular Weight (g/mol) 102.18
MDL Number MFCD00004485
InChI Key WFRBDWRZVBPBDO-UHFFFAOYSA-N
Synonym 2-methyl-2-pentanol, 2-pentanol, 2-methyl, 2-hydroxy-2-methylpentane, unii-hu2so831kp, dimethylpropylcarbinol, hu2so831kp, 1,1-dimethylbutanol, methyl-2 pentanol-2, 2-methyl-2-hydroxypentane, acmc-209m9lShow MoreShow Less
PubChem CID 11543
IUPAC Name 2-methylpentan-2-ol
SMILES CCCC(C)(C)O

Specifications

Melting Point -103°C
Boiling Point 120°C to 122°C
Flash Point 30°C (86°F)
Odor Alcohol-like
Quantity 5 g
UN Number UN2560
Beilstein 1718951
Formula Weight 102.18
Percent Purity 99%
Chemical Name or Material 2-Methyl-2-pentanol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10252249 Certificate TypeCertificate of Analysis Date02/09/2024 Catalog NumberA19090.06,A19090.14,A19090.22
Lot Number10251228 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA19090.06,A19090.14,A19090.22
Lot Number10251486 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA19090.06,A19090.14,A19090.22
Lot Number10201685 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19090.06,A19090.14,A19090.22
Lot NumberE8860C Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA19090.06,A19090.14,A19090.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methyl-2-pentanol, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.