Learn More
2-Methyl-3-nitrophenol, 96%, Thermo Scientific™
35.07 € - 355.00 €
Chemical Identifiers
CAS | 5460-31-1 |
---|---|
Molecular Formula | C7H7NO3 |
Molecular Weight (g/mol) | 153.137 |
MDL Number | MFCD00007241 |
InChI Key | GAKLFAZBKQGUBO-UHFFFAOYSA-N |
Synonym | 3-nitro-o-cresol, 2-hydroxy-6-nitrotoluene, phenol, 2-methyl-3-nitro, 2-methyl-3-nitro-phenol, 6-hydroxy-2-nitrotoluene, 3-nitro-2-methyl phenol, nitrocresol, zlchem 214, pubchem2030 |
PubChem CID | 79579 |
IUPAC Name | 2-methyl-3-nitrophenol |
SMILES | CC1=C(C=CC=C1O)[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11499893
|
Thermo Scientific Alfa Aesar
A13002.03 |
1 g |
35.07 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11409903
|
Thermo Scientific Alfa Aesar
A13002.06 |
5 g |
87.64 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11419903
|
Thermo Scientific Alfa Aesar
A13002.14 |
25 g |
355.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
5460-31-1 | |
153.137 | |
GAKLFAZBKQGUBO-UHFFFAOYSA-N | |
79579 | |
CC1=C(C=CC=C1O)[N+](=O)[O-] |
C7H7NO3 | |
MFCD00007241 | |
3-nitro-o-cresol, 2-hydroxy-6-nitrotoluene, phenol, 2-methyl-3-nitro, 2-methyl-3-nitro-phenol, 6-hydroxy-2-nitrotoluene, 3-nitro-2-methyl phenol, nitrocresol, zlchem 214, pubchem2030 | |
2-methyl-3-nitrophenol |
Specifications
5460-31-1 | |
Phenol-like | |
MFCD00007241 | |
1946057 | |
GAKLFAZBKQGUBO-UHFFFAOYSA-N | |
2-methyl-3-nitrophenol | |
79579 | |
96% |
148°C to 152°C | |
C7H7NO3 | |
UN2446 | |
3-nitro-o-cresol, 2-hydroxy-6-nitrotoluene, phenol, 2-methyl-3-nitro, 2-methyl-3-nitro-phenol, 6-hydroxy-2-nitrotoluene, 3-nitro-2-methyl phenol, nitrocresol, zlchem 214, pubchem2030 | |
CC1=C(C=CC=C1O)[N+](=O)[O-] | |
153.137 | |
153.14 | |
2-Methyl-3-nitrophenol |
Safety and Handling
GHS H Statement
H311-H302-H332-H315-H319-H335
Toxic in contact with skin.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITROCRESOLS, SOLID
missing translation for 'einecsNumber' : 226-739-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures