missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methyl-5-phenyl-3-furaldehyde, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 2-methyl-5-phenyl-3-furaldehyde, 2-methyl-5-phenyl-furan-3-carbaldehyde, 2-methyl-5-phenyl-3-furancarboxaldehyde, 3-furancarboxaldehyde,2-methyl-5-phenyl, 3-furancarboxaldehyde, 2-methyl-5-phenyl |
Formula Weight | 186.21 |
Percent Purity | 97% |
Chemical Name or Material | 2-Methyl-5-phenyl-3-furaldehyde |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'