missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-phenyl-3-furaldehyde, 97%, Thermo Scientific™

Product Code. 10511702
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 321309-42-6
Molecular Formula C12H10O2
Molecular Weight (g/mol) 186.21
MDL Number MFCD02681964
InChI Key QRBXYIYZWZGINC-UHFFFAOYSA-N
PubChem CID 2795313
IUPAC Name 2-methyl-5-phenylfuran-3-carbaldehyde
SMILES CC1=C(C=C(O1)C2=CC=CC=C2)C=O

Specifications

Assay Percent Range 97%
Quantity 1 g
Synonym 2-methyl-5-phenyl-3-furaldehyde, 2-methyl-5-phenyl-furan-3-carbaldehyde, 2-methyl-5-phenyl-3-furancarboxaldehyde, 3-furancarboxaldehyde,2-methyl-5-phenyl, 3-furancarboxaldehyde, 2-methyl-5-phenyl
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-phenyl-3-furaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'