missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethoxy)acetophenone, 97%, Thermo Scientific™

35.00 € - 117.00 €

Chemical Identifiers

CAS 1373920-71-8
Molecular Formula C10H9F3O2
Molecular Weight (g/mol) 218.175
MDL Number MFCD22201058
InChI Key XVSBANNQMCDRTC-UHFFFAOYSA-N
Synonym 2'-methyl-5'-trifluoromethoxy acetophenone, 1-2-methyl-5-trifluoromethoxy phenyl ethanone, 2-methyl-5-trifluoromethoxy acetophenone
PubChem CID 86277664
IUPAC Name 1-[2-methyl-5-(trifluoromethoxy)phenyl]ethanone
SMILES CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15478485
View Documents
Thermo Scientific Alfa Aesar
H33470.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15446277
View Documents
Thermo Scientific Alfa Aesar
H33470.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1373920-71-8
218.175
XVSBANNQMCDRTC-UHFFFAOYSA-N
86277664
CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)C
C10H9F3O2
MFCD22201058
2'-methyl-5'-trifluoromethoxy acetophenone, 1-2-methyl-5-trifluoromethoxy phenyl ethanone, 2-methyl-5-trifluoromethoxy acetophenone
1-[2-methyl-5-(trifluoromethoxy)phenyl]ethanone

Specifications

1373920-71-8
MFCD22201058
XVSBANNQMCDRTC-UHFFFAOYSA-N
1-[2-methyl-5-(trifluoromethoxy)phenyl]ethanone
86277664
97%
C10H9F3O2
2'-methyl-5'-trifluoromethoxy acetophenone, 1-2-methyl-5-trifluoromethoxy phenyl ethanone, 2-methyl-5-trifluoromethoxy acetophenone
CC1=C(C=C(C=C1)OC(F)(F)F)C(=O)C
218.175
218.17
2-Methyl-5-(trifluoromethoxy)acetophenone
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications