missing translation for 'onlineSavingsMsg'
Learn More

2-Methyl-5-(trifluoromethyl)cinnamic acid, 97%, Thermo Scientific™

Product Code. 15447945
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1017778-08-3
Molecular Formula C11H9F3O2
Molecular Weight (g/mol) 230.19
MDL Number MFCD09832288
InChI Key FJMNLFRMBNXMRQ-UHFFFAOYSA-N
Synonym 2e-3-2-methyl-5-trifluoromethyl phenyl prop-2-enoic acid, 2-methyl-5-trifluoromethyl cinnamic acid, e-3-5-trifluoromethyl-2-methylphenyl acrylic
PubChem CID 46737520
IUPAC Name (E)-3-[2-methyl-5-(trifluoromethyl)phenyl]prop-2-enoic acid
SMILES CC1=C(C=CC(O)=O)C=C(C=C1)C(F)(F)F

Specifications

Melting Point 155°C to 158°C
Quantity 250 mg
Formula Weight 230.18
Percent Purity 97%
Chemical Name or Material 2-Methyl-5-(trifluoromethyl)cinnamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'