missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester, 96%, Thermo Scientific Chemicals

85.43 € - 241.20 €

Chemical Identifiers

CAS 904326-88-1
Molecular Formula C11H18BN3O2
Molecular Weight (g/mol) 235.094
MDL Number MFCD11878345
InChI Key QDOXNCAIXITTKA-UHFFFAOYSA-N
Synonym n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid, pinacol ester, n-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid,pinacol ester, 2-methylamino pyrimidine-5-boronic acid pinacol ester, 2-methylamino pyrimidin-5-yl boronic acid pinacol ester, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl-amine, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl amine, n-m ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, n-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidin-2-amine
PubChem CID 46739641
IUPAC Name N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15471476
View Documents
Thermo Scientific Alfa Aesar
H54682.MD
250 mg
85.43 €
250mg
Please to purchase this item. Need a web account? Register with us today!
15461476
View Documents
Thermo Scientific Alfa Aesar
H54682.03
1 g
241.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Methylamino)pyrimidine-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
Specifications

Chemical Identifiers

904326-88-1
235.094
QDOXNCAIXITTKA-UHFFFAOYSA-N
46739641
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC
C11H18BN3O2
MFCD11878345
n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid, pinacol ester, n-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid,pinacol ester, 2-methylamino pyrimidine-5-boronic acid pinacol ester, 2-methylamino pyrimidin-5-yl boronic acid pinacol ester, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl-amine, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl amine, n-m ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, n-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidin-2-amine
N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

Specifications

904326-88-1
C11H18BN3O2
n-methyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid, pinacol ester, n-methyl-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, 2-methylaminopyrimidine-5-boronic acid,pinacol ester, 2-methylamino pyrimidine-5-boronic acid pinacol ester, 2-methylamino pyrimidin-5-yl boronic acid pinacol ester, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl-amine, methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyrimidin-2-yl amine, n-m ethyl-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrimidin-2-amine, n-methyl-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyrimidin-2-amine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(N=C2)NC
235.094
235.09
2-(Methylamino)pyrimidine-5-boronic acid pinacol ester
122°C to 125°C
MFCD11878345
QDOXNCAIXITTKA-UHFFFAOYSA-N
N-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine
46739641
96%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Keep cold; Store under Nitrogen

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.