missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbenzeneboronic acid 1,3-propanediol ester, 95%, Thermo Scientific™

Product Code. 15480056
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 163517-56-4
Molecular Formula C10H13BO2
Molecular Weight (g/mol) 176.022
MDL Number MFCD06659935
InChI Key UIDYFYHQHHMEKA-UHFFFAOYSA-N
Synonym 2-2-methylphenyl-1,3,2-dioxaborinane, 2-methylbenzeneboronic acid, propanediol cyclic ester, 2-methylphenylboronic acid propanediol cyclic ester, 2-o-tolyl-1,3,2-dioxaborinane, 2-2-methylphenyl 1,3,2-dioxaborinane, 1,3,2-dioxaborinane,2-2-methylphenyl, 2-methylbenzeneboronic acid 1,3-propanediol ester, 2-methylphenylboronic acid,propanediol cyclic ester, 2-methylphenylboronic acid, propanediol cyclic ester
PubChem CID 23005365
IUPAC Name 2-(2-methylphenyl)-1,3,2-dioxaborinane
SMILES B1(OCCCO1)C2=CC=CC=C2C

Specifications

Quantity 1 g
Formula Weight 176.02
Percent Purity 95%
Chemical Name or Material 2-Methylbenzeneboronic acid 1,3-propanediol ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'