Learn More
2-Methylbenzeneboronic acid, 98%, Thermo Scientific Chemicals
Description
2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Methylbenzeneboronic acid is a reagent used for Pd-catalyzed arylation using Suzuki-Miyaura cross-coupling in water, Ruthenium catalyzed direct arylation reactions, Ligand-free copper-catalyzed coupling reactions, Rhodium-catalyzed asymmetric 1,4-addition reactions. Reagent used in Preparation of chiral monophosphorus ligands in asymmetric Suzuki-Miyaura coupling reaction, Human farnesyl pyrophosphate synthase inhibitors as antitumor agents for multiple myeloma cells
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agent.
Specifications
Specifications
Melting Point | ∼162°C to 164°C |
Quantity | 5 g |
Beilstein | 2935796 |
Solubility Information | Slightly soluble in water. |
Formula Weight | 135.96 |
Percent Purity | 98% |
Chemical Name or Material | 2-Methylbenzeneboronic acid |
RUO – Research Use Only