Learn More
2-Methylbenzothiazol-5-ol, 97%, Thermo Scientific Chemicals
35.38 € - 188.94 €
Chemical Identifiers
CAS | 68867-14-1 |
---|---|
Molecular Formula | C8H7NOS |
Molecular Weight (g/mol) | 165.21 |
MDL Number | MFCD00192276 |
InChI Key | LAKVUPMDDFICNR-UHFFFAOYSA-N |
PubChem CID | 699799 |
IUPAC Name | 2-methyl-1,3-benzothiazol-5-ol |
SMILES | CC1=NC2=CC(O)=CC=C2S1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15405727
|
Thermo Scientific Alfa Aesar
H32937.03 |
1 g |
35.38 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15415727
|
Thermo Scientific Alfa Aesar
H32937.09 |
10 g |
188.94 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed as pharmaceutical intermediate.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Identifiants chimiques
68867-14-1 | |
165.21 | |
LAKVUPMDDFICNR-UHFFFAOYSA-N | |
2-methyl-1,3-benzothiazol-5-ol |
C8H7NOS | |
MFCD00192276 | |
699799 | |
CC1=NC2=CC(O)=CC=C2S1 |
Spécification
68867-14-1 | |
C8H7NOS | |
2-methylbenzo d thiazol-5-ol, 5-benzothiazolol, 2-methyl, 2-methyl-5-benzothiazolol, 2-methylbenzothiazol-5-ol, 5-benzothiazolol,2-methyl, 5-hydroxy-2-methylbenzothiazole, 2-methyl-benzothiazol-5-ol, 2-methyl-5-hydroxybenzothiazole | |
CC1=NC2=CC(O)=CC=C2S1 | |
165.21 | |
165.21 | |
2-Methylbenzothiazol-5-ol |
185°C to 187°C | |
MFCD00192276 | |
LAKVUPMDDFICNR-UHFFFAOYSA-N | |
2-methyl-1,3-benzothiazol-5-ol | |
699799 | |
97% |
Sécurité et traitement
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.