missing translation for 'onlineSavingsMsg'
Learn More

2'-Methylbiphenyl-2-carboxaldehyde, 98%, Thermo Scientific™

Product Code. 11489420
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7111-68-4
Molecular Formula C14H12O
Molecular Weight (g/mol) 196.25
MDL Number MFCD03424630
InChI Key SEEYJCMPEVOYTF-UHFFFAOYSA-N
Synonym 2-2-methylphenyl benzaldehyde, 2'-methylbiphenyl-2-carbaldehyde, 2'-methyl-1,1'-biphenyl-2-carbaldehyde, 2'-methyl-biphenyl-2-carboxaldehyde, 2'-methylbiphenyl-2-carboxaldehyde, toluol-benzaldehyd, 2-o-tolyl benzaldehyde, 2'-methyl-biphenyl-2-carbaldehyde, 2-methyl-1,1-biphenyl-2-carbaldehyde, 2'-methyl 1,1'-biphenyl-2-carbaldehyde
PubChem CID 1393189
IUPAC Name 2-(2-methylphenyl)benzaldehyde
SMILES CC1=CC=CC=C1C1=CC=CC=C1C=O

Specifications

Quantity 1 g
Sensitivity Air Sensitive
Formula Weight 196.25
Percent Purity 98%
Chemical Name or Material 2'-Methylbiphenyl-2-carboxaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'