missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methylbutane, Reagent Grade, ≥99%, Honeywell™
Reagent Grade,≥99%
50.74 € - 170.17 €
Chemical Identifiers
CAS | 78-78-4 |
---|---|
Molecular Formula | C5H12 |
Molecular Weight (g/mol) | 72.15 |
MDL Number | MFCD00009338 |
InChI Key | QWTDNUCVQCZILF-UHFFFAOYSA-N |
Synonym | isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 |
PubChem CID | 6556 |
ChEBI | CHEBI:30362 |
IUPAC Name | 2-methylbutane |
SMILES | CCC(C)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15667940
|
Honeywell Research Chemicals
M32631-500ML |
500 mL | Glass bottle |
50.74 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15637940
|
Honeywell Research Chemicals
M32631-1L |
1 L | Glass bottle |
85.45 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15647940
|
Honeywell Research Chemicals
M32631-2.5L |
2.5 L | Glass bottle |
156.41 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15657940
|
Honeywell Research Chemicals
M32631-4X1L |
4 x 1 L | Glass bottle | ||||||
Description
- ≥95% for general laboratory use.
Chemical Identifiers
78-78-4 | |
72.15 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
6556 | |
2-methylbutane |
C5H12 | |
MFCD00009338 | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
CHEBI:30362 | |
CCC(C)C |
Specifications
78-78-4 | |
Colorless | |
28°C (1013 hPa) | |
788°F | |
C5H12 | |
MFCD00009338 | |
1730723 | |
Insoluble in water | |
CCC(C)C | |
72.15 | |
2.6 (vs air) | |
72.15g/mol | |
≥99% |
-160°C | |
0.62g/mL at 25°C | |
−51°C (−59.8°F) | |
Characteristic | |
CH3CH2CH(CH3)2 | |
UN1265 | |
isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12 | |
QWTDNUCVQCZILF-UHFFFAOYSA-N | |
2-methylbutane | |
6556 | |
CHEBI:30362 | |
11.17psi (20°C) | |
2-Methylbutane |
Safety and Handling
P210-P261-P273-P301 + P310-P331
missing translation for 'dotInformation' : class 3 - PG 1 - Pentanes
missing translation for 'rtecsNumber' : 201-142-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 2-Methylbutane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 1
- Aspiration toxicity Category 1
- Specific target organ toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
- H224-Extremely flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H336-May cause drowsiness or dizziness.
- H411-Toxic to aquatic life with long lasting effects.
- EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.