missing translation for 'onlineSavingsMsg'
Learn More
2-Methylbutane, 99+%, Thermo Scientific Chemicals Product Code.: 11410500

2-Methylbutane, 99+%, Thermo Scientific Chemicals

Product Code. 11410500
4 x 500 mL
Click to view available options
Quantity:
500 mL
4 x 500 mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11410500

Brand: Thermo Scientific Alfa Aesar 019387.AY

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

2-Methylbutane is used as a chemical intermediate. It acts as a blowing agent for polystyrene and gasoline additive. It is used as a solvent of polyethylene and involved in the preparation of polystyrene foam and polyurethane foam. Further, it is used in a closed loop in geothermal power production to operate turbines. In addition to this, it is used to freeze biological samples like tissues in place of dry ice for cryosectioning in histology.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12Show MoreShow Less
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Density 0.62
Boiling Point 30°C
Flash Point -57°C (-70°F)
Assay Percent Range 99+%
Refractive Index 1.354
Quantity 4 x 500 mL
Solubility Information Miscible with water,hydrocarbons,oils,alcohol and ether.
Formula Weight 72.14
Physical Form Liquid
Chemical Name or Material 2-Methylbutane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY12K037 Certificate TypeCertificate of Analysis Date25/02/2025 Catalog Number019387.AP,019387.AY
Lot NumberZ23K008 Certificate TypeCertificate of Analysis Date23/01/2025 Catalog Number019387.AP,019387.AY
Lot NumberX22K028 Certificate TypeCertificate of Analysis Date30/10/2024 Catalog Number019387.AP,019387.AY
Lot NumberX03K044 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog Number019387.AP,019387.AY
Lot NumberU05K013 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog Number019387.AP,019387.AY
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylbutane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H411-Toxic to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Isopentane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methylbutane, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.