missing translation for 'onlineSavingsMsg'
Learn More

2-Methylbutane, Puriss p.a., ≥99.5% (GC), Honeywell Riedel-de Haën™

Product Code. 15665880
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Puriss. p.a.,≥99.5% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-78-4
Molecular Formula C5H12
Molecular Weight (g/mol) 72.15
MDL Number MFCD00009338
InChI Key QWTDNUCVQCZILF-UHFFFAOYSA-N
Synonym isopentane, isoamylhydride, butane, 2-methyl, dimethylethylmethane, ethyldimethylmethane, 1,1,2-trimethylethane, iso-pentane, 1,1-dimethylpropane, butanes, iso-c5h12
PubChem CID 6556
ChEBI CHEBI:30362
IUPAC Name 2-methylbutane
SMILES CCC(C)C

Specifications

Melting Point -160°C
Density 0.62g/mL at 25°C
Boiling Point 28°C
Flash Point −51°C (−59.8°F)
Ignition Point 788°F
Packaging Glass bottle
Linear Formula CH3CH2CH(CH3)2
Refractive Index 1.354
Quantity 2.5 L
UN Number UN1265
Beilstein 1730723
Vapor Density 2.6 (vs air)
Formula Weight 72.15g/mol
Vapor Pressure 11.17psi (20°C)
Percent Purity ≥99.5% (GC)
Chemical Name or Material 2-Methylbutane
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.