missing translation for 'onlineSavingsMsg'
Learn More
2-Methylcyclohexanol, 99%, mixture of cis and trans, Thermo Scientific Chemicals Product Code.: 10522191

2-Methylcyclohexanol, 99%, mixture of cis and trans, Thermo Scientific Chemicals

Product Code. 10522191
250 g
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
This item is not returnable. View return policy

Product Code. 10522191

Brand: Thermo Scientific Acros 166892500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-59-5
Molecular Formula C7H14O
Molecular Weight (g/mol) 114.19
MDL Number MFCD00001443
InChI Key NDVWOBYBJYUSMF-UHFFFAOYSA-N
Synonym 2-methylcyclohexanol, cyclohexanol, 2-methyl, o-methylcyclohexanol, 1-methyl-2-cyclohexanol, 2-methyl-1-cyclohexanol, 2-methylcyclohexyl alcohol, cyclohexanol, o-methyl, cis-2-methylcyclohexanol, 2-methyl-cyclohexanol, 2-methylcyclohexanol, mixed isomers
PubChem CID 11418
IUPAC Name 2-methylcyclohexan-1-ol
SMILES CC1CCCCC1O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -38.0°C
Color Colorless
Density 0.9300g/mL
Boiling Point 163.0°C to 166.0°C
Flash Point 58°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H10OH
Refractive Index 1.4615 to 1.4635
Quantity 250 g
Beilstein 06, 11
Fieser 08,303
Specific Gravity 0.93
Solubility Information Solubility in water: slightly soluble
Formula Weight 114.19
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylcyclohexanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methylcyclohexanol, 99%, mixture of cis and trans, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.