missing translation for 'onlineSavingsMsg'
Learn More

2-Methylfuran, 99%, stabilized, Thermo Scientific Chemicals

Product Code. 10081140
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-22-5, 123-31-9
MDL Number MFCD00003248
InChI Key VQKFNUFAXTZWDK-UHFFFAOYSA-N
Synonym methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
PubChem CID 10797
IUPAC Name 2-methylfuran
SMILES CC1=CC=CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -88.7°C
Color Yellow
Density 0.9100g/mL
Boiling Point 63.0°C to 66.0°C
Flash Point −26°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C5H6O
Refractive Index 1.4320 to 1.4340
Quantity 5 mL
Beilstein 17, 36
Fieser 01,682; 14,209
Specific Gravity 0.91
Solubility Information Solubility in water: 0.3g/100mL(20 c). Other solubilities: miscible with most common organic solvents
Stabilizer 0.01% hydroquinone
Molecular Weight (g/mol) 82.1
Viscosity 4 mPa.s (25°C)
Formula Weight 82.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylfuran, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'