missing translation for 'onlineSavingsMsg'
Learn More

2-Methylfuran, 99%, stabilized

Product Code. 10552521 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
5mL
This item is not returnable. View return policy

Product Code. 10552521

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 534-22-5 | C5H6O | 82.1 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 534-22-5, 123-31-9
MDL Number MFCD00003248
InChI Key VQKFNUFAXTZWDK-UHFFFAOYSA-N
Synonym methylfuran, sylvan, 2-methyl furan, silvan, furan, 2-methyl, 5-methylfuran, alpha-methylfuran, furan, methyl, silvan czech, .alpha.-methylfuran
PubChem CID 10797
IUPAC Name 2-methylfuran
SMILES CC1=CC=CO1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -88.7°C
Color Yellow
Density 0.9100g/mL
Boiling Point 63.0°C to 66.0°C
Flash Point −26°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C5H6O
Refractive Index 1.4320 to 1.4340
Quantity 1 L
Beilstein 17, 36
Fieser 01,682; 14,209
Specific Gravity 0.91
Stabilizer 0.01% hydroquinone
Molecular Weight (g/mol) 82.102
Formula Weight 82.1
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Methylfuran, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.