missing translation for 'onlineSavingsMsg'
Learn More

2-Methylheptane, 99%, Thermo Scientific Chemicals

Product Code. 11366649
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

2-methylheptane is a gas detector and it is used in RD DIN40 respirator mask with gas filter type A.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-methylheptane is a gas detector and it is used in RD DIN40 respirator mask with gas filter type A.

Solubility
Soluble in alcohol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible material is strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-27-8
Molecular Formula C8H18
Molecular Weight (g/mol) 114.232
MDL Number MFCD00009518
InChI Key JVSWJIKNEAIKJW-UHFFFAOYSA-N
Synonym heptane, 2-methyl, methylheptane, 2-methyl-heptane, unii-yu6su8ccvb, yu6su8ccvb, hexane, dimethyl, dimethylhexane, acmc-1avkm, 2-methylheptane, ch3 2ch ch2 4ch3
PubChem CID 11594
IUPAC Name 2-methylheptane
SMILES CCCCCC(C)C

Specifications

Density 0.698
Boiling Point 116°C
Flash Point 4°C (40°F)
Linear Formula (CH3)2CHCH2CH2CH2CH2CH3
Refractive Index 1.395
Quantity 5 g
UN Number UN1262
Beilstein 1696862
Solubility Information Soluble in alcohol. Insoluble in water.
Formula Weight 114.23
Percent Purity 99%
Chemical Name or Material 2-Methylheptane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'