missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazo[1,2-a]pyridine-3-carboxylic acid, 97%, Thermo Scientific™

41.90 € - 295.00 €

Chemical Identifiers

CAS 21801-79-6
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.18
MDL Number MFCD00244615
InChI Key FBIUGCLQMKPURJ-UHFFFAOYSA-N
Synonym 2-methylimidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl, 2-methyl-4-hydroimidazo 1,2-a pyridine-3-carboxylic acid, pubchem16305, imidazo 1, 2-methyl, 2-methylimidazo 1,2-a nicotinic acid, 3-carboxy-2-methylimidazo 1,2-a pyridine
PubChem CID 89054
IUPAC Name 2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
SMILES CC1=C(N2C=CC=CC2=N1)C(O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10147752
View Documents Promotion Details
Thermo Scientific Maybridge
CC21701CB
250 mg
41.90 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10126312
View Documents Promotion Details
Thermo Scientific Maybridge
CC21701DA
1 g
90.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
10712017
View Documents Promotion Details
Thermo Scientific Maybridge
CC21701DE
5 g
295.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

21801-79-6
176.18
FBIUGCLQMKPURJ-UHFFFAOYSA-N
89054
CC1=C(N2C=CC=CC2=N1)C(O)=O
C9H8N2O2
MFCD00244615
2-methylimidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl, 2-methyl-4-hydroimidazo 1,2-a pyridine-3-carboxylic acid, pubchem16305, imidazo 1, 2-methyl, 2-methylimidazo 1,2-a nicotinic acid, 3-carboxy-2-methylimidazo 1,2-a pyridine
2-methylimidazo[1,2-a]pyridine-3-carboxylic acid

Specifications

21801-79-6
100.0
Amber glass bottle
MFCD00244615
FBIUGCLQMKPURJ-UHFFFAOYSA-N
2-methylimidazo[1,2-a]pyridine-3-carboxylic acid
89054
97%
96.5
97%
C9H8N2O2
2-methylimidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl-imidazo 1,2-a pyridine-3-carboxylic acid, imidazo 1,2-a pyridine-3-carboxylic acid, 2-methyl, 2-methyl-4-hydroimidazo 1,2-a pyridine-3-carboxylic acid, pubchem16305, imidazo 1, 2-methyl, 2-methylimidazo 1,2-a nicotinic acid, 3-carboxy-2-methylimidazo 1,2-a pyridine
CC1=C(N2C=CC=CC2=N1)C(O)=O
176.18
176.17
2-Methylimidazo[1, 2-a]pyridine-3-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications