missing translation for 'onlineSavingsMsg'
Learn More

2-Methylimidazole, 99%, Thermo Scientific Chemicals

Product Code. 10346530
Click to view available options
Quantity:
100 g
500 g
1 kg
Packaging:
Plastic bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 693-98-1
Molecular Formula C4H6N2
Molecular Weight (g/mol) 82.11
MDL Number MFCD00005190
InChI Key LXBGSDVWAMZHDD-UHFFFAOYSA-N
Synonym 2-methylimidazole, 1h-imidazole, 2-methyl, imidazole, 2-methyl, 2-methyl imidazole, 2-methylglyoxaline, unii-t0049z45lz, ccris 2459, dsstox_cid_2107, dsstox_rid_76489, dsstox_gsid_22107
PubChem CID 12749
IUPAC Name 2-methyl-1H-imidazole
SMILES CC1=NC=CN1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 142.0°C to 145.0°C
Color White to Yellow
Boiling Point 270.0°C
Flash Point 155°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Quantity 500 g
Beilstein 23, 65
Solubility Information Solubility in water: 267g/L (20°C)
Formula Weight 82.1
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 2-Methylimidazole, 99%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Methylimidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H360-May damage fertility or the unborn child.
  • H351-Suspected of causing cancer.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'