missing translation for 'onlineSavingsMsg'
Learn More
2-Methylindole, 98+%, Thermo Scientific Chemicals Product Code.: 11449662

2-Methylindole, 98+%, Thermo Scientific Chemicals

Product Code. 11449662
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11449662

Brand: Thermo Scientific Alfa Aesar A10764.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Methylindole is used as a reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction, Friedel-Crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, Michael addition reactions and in synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Methylindole is used as a reactant for regioselective synthesis of oxopyrrolidine analogs via iodine-catalyzed Markovnikov addition reaction, Friedel-Crafts alkylation reactions, preparation of tryptophan dioxygenase inhibitors pyridyl-ethenyl-indoles as potential anticancer immunomodulators, Michael addition reactions and in synthesis of cyclooxygenase-1 (COX-1)/cyclooxygenase-2 (COX-2) inhibitors.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-20-5
Molecular Formula C9H9N
Molecular Weight (g/mol) 131.178
MDL Number MFCD00005616
InChI Key BHNHHSOHWZKFOX-UHFFFAOYSA-N
Synonym 2-methylindole, 1h-indole, 2-methyl, indole, 2-methyl, 2-methyl indole, unii-i7cn58827i, 2-methylindol, methylketole, methyl indole, 2-methyl-indole, pubchem7504
PubChem CID 7224
ChEBI CHEBI:49402
IUPAC Name 2-methyl-1H-indole
SMILES CC1=CC2=CC=CC=C2N1

Specifications

Melting Point 56°C to 60°C
Density 1.07
Boiling Point 272°C to 273°C
Flash Point 141°C (285°F)
Quantity 50 g
Beilstein 109781
Sensitivity Air and light sensitive
Solubility Information Insoluble in water.
Formula Weight 131.18
Percent Purity ≥98%
Chemical Name or Material 2-Methylindole
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10244667 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog NumberA10764.18,A10764.30,A10764.0B
Lot Number10237429 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA10764.18,A10764.30,A10764.0B
Lot Number10224286 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10764.18,A10764.30,A10764.0B
Lot Number10117352 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10764.18,A10764.30,A10764.0B
Lot Number10231042 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA10764.18,A10764.30,A10764.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Methylindole, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.